Learn More
Grainger 6-Inch Work Boot M 11 Black PR
Catalog No. 502766512
Encompass_Preferred
The Grainger 16V677 is a Dayton 1/3 HP 115/230V AC electric motor. It is a single phase, open drip proof motor with a 56C frame and a 1075 RPM operating speed. The motor has a 1/2" diameter shaft and includes a capacitor. It can provide 0.25 horsepower output and is thermally protected. The 16V677 is designed for general purpose applications.
Catalog No. 50-276-6512 Supplier Grainger Supplier No. 16V677
The Grainger 16V677 is a Dayton 1/3 HP 115/230V AC electric motor. It is a single phase, open drip proof motor with a 56C frame and a 1075 RPM operating speed. The motor has a 1/2" diameter shaft and includes a capacitor. It can provide 0.25 horsepower output and is thermally protected. The 16V677 is designed for general purpose applications.