Learn More
Grainger 6-Inch Work Boot M 12 Black PR
Catalog No. 502766508
Encompass_Preferred
The Grainger 16V679 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with an aluminum end shield. This motor can provide 0.37 kW of output power. It has Class F insulation and a NEMA standard 56C frame. The 16V679 is built for general purpose applications.
Catalog No. 50-276-6508 Supplier Grainger Supplier No. 16V679
The Grainger 16V679 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with an aluminum end shield. This motor can provide 0.37 kW of output power. It has Class F insulation and a NEMA standard 56C frame. The 16V679 is built for general purpose applications.