Learn More
Grainger 6-Inch Work Boot M 6 1/2 Black PR
Catalog No. 502766523
Encompass_Preferred
The Norton PSA Sanding Disc Roll is a 5 inch diameter sanding disc made from aluminum oxide grain on a flexible paper backing. It has pressure sensitive adhesive for quick attachment and removal. The disc has an open coat design and is intended for light sanding, smoothing, and finishing on wood, metal, plastic, drywall, and paint. It comes in a roll with 25 discs. The grit sizes available are 80, 100, 150, and 220.
Catalog No. 50-276-6523 Supplier Grainger Supplier No. 16V668
The Norton PSA Sanding Disc Roll is a 5 inch diameter sanding disc made from aluminum oxide grain on a flexible paper backing. It has pressure sensitive adhesive for quick attachment and removal. The disc has an open coat design and is intended for light sanding, smoothing, and finishing on wood, metal, plastic, drywall, and paint. It comes in a roll with 25 discs. The grit sizes available are 80, 100, 150, and 220.