Learn More
Sigma Aldrich 6-Iodo-3,4-dihydro-2H-pyrano[3,2-b]pyridin-8-ol
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Cis-1,4-hexadiene can be synthesized from the cobalt-catalyzed reaction between butadiene and ethylene
- The reaction of this hydrocarbon with fe(co)5 affords the trans-1,3-hexadiene-iron tricarbonyl complex (i), plus a small amount of the trans,trans-2,4-hexadiene complex(ii)
- Mechanism of the conversion of cis-1,4-hexadiene to trans-2-methyl-1,3-pentadiene, via rearrangement in the presence of a nickel catalyst has been investigated
Specifications
Specifications
| Molecular Formula | C8H8INO2 |
| Linear Formula | C8H8INO2 |
| MDL Number | MFCD20487127 |
| Quantity | 5 mL |
| Molecular Weight (g/mol) | 277.06 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.