Learn More
TARGETMOL CHEMICALS INC 6-IODOPRAVADOLINE 25MG
SDP
Catalog No. 502581306
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 6-Iodopravadoline (AM630) is an antagonist of CB2 (Ki 31.2 nM). And it shows 165-fold selectivity more than CB1 receptors. purity: 99%
Catalog No. 50-258-1306 Supplier TARGETMOL CHEMICALS INC Supplier No. T1420425MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 6-Iodopravadoline (AM630) is an antagonist of CB2 (Ki 31.2 nM). And it shows 165-fold selectivity more than CB1 receptors. purity: 99%