Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-(Isopropylthio)imidazo[2,1-b]thiazole-5-carbaldehyde

Catalog No. OTV0009331G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0009331G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0009331G Supplier Sigma Aldrich Supplier No. OTV0009331G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Structures of 2-phenylethanethiol and its 1:1 water clusters have been studied using resonant two-photon ionization spectroscopy.2-phenylethanethiol has been used:
  • In preparation of biicosahedral au (25) clusters protected with various types of thiol ligands
  • As derivatization reagent for phosphorylated sequences of peptides, for enhancing ionization efficiency in matrix-assisted laser desorption/ionization mass spectrometry

Specifications

CAS 338976-44-6
Molecular Formula C9H10N2OS2
Linear Formula C9H10N2OS2
MDL Number MFCD00139187
Quantity 10 g
Molecular Weight (g/mol) 226.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.