Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-isothiocyanato-fluorescein | 3012-71-3 | MFCD00041838 | 90.0% | 389.38 g·mol⁻¹ | C21H11NO5S | 250 MG
SDP

Catalog No. 5000284795
Encompass_Preferred

6-Isothiocyanato-Fluorescein (FITC isomer II) is an amine-reactive fluorescent labeling reagent used to derivatize primary amines on proteins, peptides, and other biomolecules for fluorescence detection and conjugation workflows. It is supplied as a solid and reacts via its isothiocyanate group to form stable thiourea linkages.

  • Amine-reactive isothiocyanate group for stable thiourea bond formation.
  • Suitable for labeling proteins, peptides, and small molecules for fluorescence detection.
  • Offered as a 250 MG solid convenient for multiple conjugation reactions.
  • Typical purity approximately 90.0% by supplier specification.
  • Molecular formula C21H11NO5S and molecular weight 389.38 g·mol⁻¹.
  • CAS number 3012-71-3 for unambiguous chemical identification.

Catalog No. 50-002-84795 Supplier Medchemexpress LLC Supplier No. HYW589849250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Isothiocyanato-Fluorescein (FITC isomer II) is an amine-reactive fluorescent labeling reagent used to derivatize primary amines on proteins, peptides, and other biomolecules for fluorescence detection and conjugation workflows. It is supplied as a solid and reacts via its isothiocyanate group to form stable thiourea linkages.

  • Amine-reactive isothiocyanate group for stable thiourea bond formation.
  • Suitable for labeling proteins, peptides, and small molecules for fluorescence detection.
  • Offered as a 250 MG solid convenient for multiple conjugation reactions.
  • Typical purity approximately 90.0% by supplier specification.
  • Molecular formula C21H11NO5S and molecular weight 389.38 g·mol⁻¹.
  • CAS number 3012-71-3 for unambiguous chemical identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.