Learn More
AdipoGen 6-JOE SE
SDP
Catalog No. 502561471
Encompass_Preferred
Chemical. CAS 113394-23-3. Formula C27H17Cl2NO11. MW 602.34. Synthetic. Amine-reactive fluorescent probe. A conventional fluorophore predominantly used in automated DNA sequencing. It is also used to label nucleotides. Characterized by an intermediate-wavelength spectra, high quantum yield, and low pH sensitivity in the physiological range. Has a similar absorption and emission spectra to those of rhodamine 6G. Spectral Data lambdaex 520nm, lambdaem550 nm in Buffer pH 12 lambdaex 520nm, lambdaem 548nm in DMSO.
Catalog No. 50-256-1471 Supplier AdipoGen Supplier No. CDXC0098M005
Chemical. CAS 113394-23-3. Formula C27H17Cl2NO11. MW 602.34. Synthetic. Amine-reactive fluorescent probe. A conventional fluorophore predominantly used in automated DNA sequencing. It is also used to label nucleotides. Characterized by an intermediate-wavelength spectra, high quantum yield, and low pH sensitivity in the physiological range. Has a similar absorption and emission spectra to those of rhodamine 6G. Spectral Data lambdaex 520nm, lambdaem550 nm in Buffer pH 12 lambdaex 520nm, lambdaem 548nm in DMSO.