Learn More
Enzo Life Sciences 6-Ketoprostaglandin F1alpha (1mg). CAS: 58962-34-8
SDP
Supplier:  Enzo Life Sciences BMLPG0170001
Encompass_Preferred
Biosynthesis via hydrolysis of prostacyclin. A reliable parameter for measurement of prostacyclin. Alternative name: 6-Keto-PGF1alpha. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1420