Learn More
Medchemexpress LLC 6-Maleimidocaproic acid-PFP ester | 692739-25-6 | C16H12F5NO4 | 250 MG
SDP
Catalog No. 5000352723
Encompass_Preferred
6-Maleimidocaproic acid-PFP ester is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52723 Supplier Medchemexpress LLC Supplier No. 250MGHY140990
6-Maleimidocaproic acid-PFP ester is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins