Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Maleimidohexanoic acid

Catalog No. AA4638414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4638414 25 g
AA4638406 5 g
AA4638422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4638414 Supplier Thermo Scientific Chemicals Supplier No. 04638414
Only null left
Add to Cart

CAS: 55750-53-3 | C10H13NO4 | 211.217 g/mol

6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55750-53-3
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.217
MDL Number MFCD00043140
InChI Key WOJKKJKETHYEAC-UHFFFAOYSA-N
Synonym 6-maleimidocaproic acid, 6-maleimidohexanoic acid, 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoic acid, 6-maleimidocaproicacid, 1h-pyrrole-1-hexanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 5-cooh, n-maleoyl-6-aminocaproic acid, n-maleoyl-6-aminohexanoic acid, 1h-pyrrole-1-hexanoicacid, 2,5-dihydro-2,5-dioxo, 6-2,5-dioxopyrrol-1-yl hexanoic acid
PubChem CID 573683
IUPAC Name 6-(2,5-dioxopyrrol-1-yl)hexanoic acid
SMILES C1=CC(=O)N(C1=O)CCCCCC(=O)O

Specifications

Color White to Yellow
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,methanol,ethanol,dimethyl sulfoxide. Insoluble in ether.
Formula Weight 211.22
Physical Form Powder
Chemical Name or Material 6-Maleimidohexanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.