Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Maleimidohexanoic acid

Catalog No. 502891508
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-1508 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-289-1508 Supplier Sigma Aldrich Supplier No. 75584225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

6-Maleimidohexanoic acid
TRUSTED_SUSTAINABILITY

Specifications

CAS 55750-53-3
Melting Point 87°C to 92°C
Molecular Formula C10H13NO4
Linear Formula C10H13NO4
MDL Number MFCD00043140
Quantity 25 g
Synonym 2,5-Dihydro-2,5-dioxo-1 H-pyrrole-1-hexanoic acid; 6-Maleimide hexanoic acid; MCA; Maleimidocaproic acid; 6-Maleimidocaproic acid; N-Maleoyl-6-aminocaproic acid; N-(5-Carboxypentyl)maleimide
Molecular Weight (g/mol) 211.21
Percent Purity 90% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.