Learn More
Medchemexpress LLC 6-maleimidohexanoic acid N-hydroxysuccinimide ester | 55750-63-5 | MFCD00043043 | 100.0% | 308.29 g/mol | C14H16N2O6 | 500 MG
SDP
Catalog No. 5000202939
Encompass_Preferred
6-Maleimidohexanoic acid N-hydroxysuccinimide ester is a heterobifunctional cross-linking reagent containing maleimide and NHS ester reactive groups that enable selective conjugation between thiol and amine functionalities for preparation of hapten and enzyme immunoconjugates.
  • Heterobifunctional reagent: maleimide and NHS ester groups.
  • Reacts with thiols and primary amines for targeted conjugation.
  • Molecular weight 308.29 g/mol; formula C14H16N2O6.
  • High purity suitable for analytical and preparative applications.
  • Available in multiple pack sizes, including 500 MG.
  • Store sealed, away from moisture and light; store solid at -20°C.
Catalog No. 50-002-02939 Supplier Medchemexpress LLC Supplier No. HY78961500MG
6-Maleimidohexanoic acid N-hydroxysuccinimide ester is a heterobifunctional cross-linking reagent containing maleimide and NHS ester reactive groups that enable selective conjugation between thiol and amine functionalities for preparation of hapten and enzyme immunoconjugates.
  • Heterobifunctional reagent: maleimide and NHS ester groups.
  • Reacts with thiols and primary amines for targeted conjugation.
  • Molecular weight 308.29 g/mol; formula C14H16N2O6.
  • High purity suitable for analytical and preparative applications.
  • Available in multiple pack sizes, including 500 MG.
  • Store sealed, away from moisture and light; store solid at -20°C.