Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Mercapto-1-hexanol

Catalog No. 45108825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-108-825ML 25 mL
45-108-85ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-108-825ML Supplier Sigma Aldrich Supplier No. 45108825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

6-mecapto-1-hexanol (MCH) forms a self-assembled monolayer (SAM) with identically represented C6 spacer. MCH is a tool for surface coating that enables blocking of active sites and non-specific absorption.MCH is coated on a piezoelectric crystal which can be used for the fabrication of chemical sensor to differentiate between cheese varieties. Self assembled monolayers of MCH can be used on gold surfaces for bio-sensing applications. It can also be used as a monolayer for the surface modification of gold-polyaniline (Au-PANI) nanocomposites for the development of aptamer based biosensors (aptasensors).

Specifications

CAS 1633-78-9
Density 0.981 g/mL (at 25°C)
Boiling Point 225°C
Molecular Formula C6H14OS
Refractive Index n20/D 1.486 (literature)
Linear Formula HS(CH2)6 OH
Quantity 25 mL
MDL Number MFCD00068552
Synonym 6-Hydroxy-1-hexanethiol; 6-MCH; 6-Mercaptohexan-1-ol; 6-Mercaptohexanol; MCH
Molecular Weight (g/mol) 134.24
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.