Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Mercaptohexanoic acid

Catalog No. 6749741G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-497-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-497-41G Supplier Sigma Aldrich Supplier No. 6749741G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

6-Mercaptohexanoic acid (MHA) forms a self-assembled monolayer (SAM) that is used to cap a variety of materials like gold nanoparticles (AuNPs) and indium phosphide quantum dots (InPQDs). It is majorly used in immobilizing the surface molecules, which enhance the physio-chemical and electrical properties.Used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles. This compound is used in self-assembly to produce hydrophilic SAMs. The resulting monolayers which are terminated with carboxylic acids can be further functionalized with various amines and alcohols to introduce more complex end groups. Can also be used to control the rate of aggregation of gold colloidal nanoparticles using pH.

Specifications

CAS 17689-17-7
Density 1.0715 g/mL (at 25°C)
Molecular Formula C6H12O2S
Refractive Index n20/D 1.4846
Linear Formula C6H12O2S
Quantity 1 g
MDL Number MFCD08705278
Synonym MHA
Molecular Weight (g/mol) 148.22
Percent Purity 90%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.