Learn More
Medchemexpress LLC HY-13677 100mg , 6-Mercaptopurine CAS:50-44-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13677100MG
Encompass_Preferred
HY-13677 100mg 6-Mercaptopurine CAS: 50-44-2 6-Mercaptopurine is a purine analogue which acts as an antagonist of the endogenous purines and has been widely used as antileukemic agent and immunosuppressive drug. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0797