Learn More
Research Products International Corp 6-Mercaptopurine, 10 Grams
SDP
Catalog No. 50213445
Encompass_Preferred
Analog of the natural purines hypoxanthine and adenine. Antineoplastic agent which acts as an antimetabolite. Toxicity: LD50 < 364 mg/kg.
WARNING! This product can expose you to 6-Mercaptopurine hydrate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-445 Supplier Research Products International Corp Supplier No. M2202010.0
Analog of the natural purines hypoxanthine and adenine. Antineoplastic agent which acts as an antimetabolite. Toxicity: LD50 < 364 mg/kg.
WARNING! This product can expose you to 6-Mercaptopurine hydrate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.