Learn More
Medchemexpress LLC 6-Mercaptopurine-13C2,15N | 1190008-04-8 | 96.0% | 155.16 | 1 MG
SDP
Catalog No. 5000348106
Encompass_Preferred
6-Mercaptopurine-13C2,15N is a 13C- and 15N-labeled version of 6-Mercaptopurine, a purine analogue that functions as an antagonist of endogenous purines and has been widely utilized as an antileukemic and immunosuppressive agent. This labeled compound is intended for research use only and is not sold to patients. Stable heavy isotopes like those in this product have been incorporated into drug molecules for quantitation during drug development, with deuteration specifically noted for its potential impact on pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-48106 Supplier Medchemexpress LLC Supplier No. HY13677S11MG
Add to Cart
Add to Cart
6-Mercaptopurine-13C2,15N is a 13C- and 15N-labeled version of 6-Mercaptopurine, a purine analogue that functions as an antagonist of endogenous purines and has been widely utilized as an antileukemic and immunosuppressive agent. This labeled compound is intended for research use only and is not sold to patients. Stable heavy isotopes like those in this product have been incorporated into drug molecules for quantitation during drug development, with deuteration specifically noted for its potential impact on pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS