Learn More
Selleck Chemical LLC 6-Mercaptopurine (6-MP) Monohydrate S4504-200mg
SDP
Catalog No. 503108857
Encompass_Preferred
6-Mercaptopurine (6-MP) Monohydrate is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA
Catalog No. 50-310-8857 Supplier Selleck Chemical LLC Supplier No. S4504200MG
6-Mercaptopurine (6-MP) Monohydrate is a widely used antileukemic agent and immunosuppressive drug that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA