Learn More
Alkali Scientific 6 Mercaptopurine, 5 G
SDP
Catalog No. 5000660295
Encompass_Preferred
Mercaptopurine is a purine analog used extensively in research on cancer treatment and immune system modulation. This 5g bottle of mercaptopurine is often used in laboratory experiments to study the effects of purine metabolism inhibition on cellular growth and DNA synthesis. It is most commonly used in the treatment of leukemia and lymphoma, as it disrupts the production of purine nucleotides, essential building blocks for DNA replication. The product is also used in studies involving organ transplantation and autoimmune disorders, where its immunosuppressive properties can be explored. This reagent is critical for understanding the mechanisms underlying chemotherapy and immunosuppressive therapy.
Catalog No. 50-006-60295 Supplier Alkali Scientific Supplier No. ERC002CP
Mercaptopurine is a purine analog used extensively in research on cancer treatment and immune system modulation. This 5g bottle of mercaptopurine is often used in laboratory experiments to study the effects of purine metabolism inhibition on cellular growth and DNA synthesis. It is most commonly used in the treatment of leukemia and lymphoma, as it disrupts the production of purine nucleotides, essential building blocks for DNA replication. The product is also used in studies involving organ transplantation and autoimmune disorders, where its immunosuppressive properties can be explored. This reagent is critical for understanding the mechanisms underlying chemotherapy and immunosuppressive therapy.