Learn More
Medchemexpress LLC 6-mercaptopurine hydrate | 6112-76-1 | MFCD03854445 | 99.9% | 170.19 g/mol | C5H6N4OS | 5 MG
SDP
Catalog No. 5000221854
Encompass_Preferred
6-mercaptopurine hydrate is the hydrated form of the purine analogue 6-mercaptopurine, used in research as an inhibitor of purine metabolism and as a reference compound in biochemical and pharmacological assays.
  • Hydrated purine analogue; CAS 6112-76-1.
  • Molecular formula C5H6N4OS and molecular weight 170.19 g/mol.
  • High purity (99.9% by HPLC) suitable for analytical use.
  • Solid monohydrate form with consistent batch characterization.
  • Commonly used in in vitro and in vivo pharmacology and biochemical assays.
  • Supplied in small research pack sizes for laboratory applications.
Catalog No. 50-002-21854 Supplier Medchemexpress LLC Supplier No. HY13677A5MG
6-mercaptopurine hydrate is the hydrated form of the purine analogue 6-mercaptopurine, used in research as an inhibitor of purine metabolism and as a reference compound in biochemical and pharmacological assays.
  • Hydrated purine analogue; CAS 6112-76-1.
  • Molecular formula C5H6N4OS and molecular weight 170.19 g/mol.
  • High purity (99.9% by HPLC) suitable for analytical use.
  • Solid monohydrate form with consistent batch characterization.
  • Commonly used in in vitro and in vivo pharmacology and biochemical assays.
  • Supplied in small research pack sizes for laboratory applications.