Learn More
6-Mercaptopurine monohydrate, 98%
Description
6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.
Solubility
Soluble in dilute solutions of alkali hydroxides, hot alcohol, and 1 N NaOH (50 mg/ml). Insoluble in water.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | ∼315°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 4012091 |
| Merck Index | 14,5871 |
| Solubility Information | Soluble in dilute solutions of alkali hydroxides,hot alcohol,and 1 N NaOH (50mg/ml). Insoluble in water. |
| Formula Weight | 170.19 (152.17 Anhydrous) |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Mercaptopurine monohydrate |
Safety and Handling
| Hazard Category | H302-H315-H319-H335-H341-H361 |
| Hazard Statement | GHS H Statement H341-H361-H302 Suspected of causing genetic defects. Suspected of damaging fertility or the unborn child. Harmful if swallowed. |
| Precautionary Statement | P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 200-037-4 |
| RTECSNumber | UP0400000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.