Learn More
Cayman Chemical 6-Mercaptopurine (hydrate), Purity: ≥98%, CAS: 6112-76-1, 10 g
SDP
Catalog No. 502994663
Encompass_Preferred
An inhibitor of purine synthesis and interconversion; rapidly converted to 6-mercaptopurine ribonucleoside-5'-monophosphate, which inhibits PRPP amidotransferase, the conversion of IMP to adenylosuccinic acid and xanthylic acid, and blocks AMP formation in vitro; inhibits growth of sarcoma 180, adenocarcinoma E 0771, and adenocarcinoma 755 tumors and reduces the size of leukemia L1210 subcutaneous growths in mice when administered at a dose of 30 mg/kg; decreases delayed-type hypersensitivity and thyroid inflammation in a guinea pig model of thyroiditis when administered pre- or post-disease onset
Catalog No. 50-299-4663 Supplier Cayman Chemical Supplier No. 2367510 G
An inhibitor of purine synthesis and interconversion; rapidly converted to 6-mercaptopurine ribonucleoside-5'-monophosphate, which inhibits PRPP amidotransferase, the conversion of IMP to adenylosuccinic acid and xanthylic acid, and blocks AMP formation in vitro; inhibits growth of sarcoma 180, adenocarcinoma E 0771, and adenocarcinoma 755 tumors and reduces the size of leukemia L1210 subcutaneous growths in mice when administered at a dose of 30 mg/kg; decreases delayed-type hypersensitivity and thyroid inflammation in a guinea pig model of thyroiditis when administered pre- or post-disease onset