Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Catalog No. CDS002721 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002721 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002721 Supplier Sigma Aldrich Supplier No. CDS002721100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3-dihydro-2,2-dimethyl-7-benzofuranol is a 7-phenol metabolite formed during the hydrolysis of carbofuran
  • 2,3-dihydro-2,2-dimethyl-7-benzofuranol (ddbfo) reacts with ti(oipr)4 or zr(oet)4 to yield dimers of [ti(micro-ddbfo)2(ddbfo)6] and [zr(ddbfo)3(etoh)(micro-eto)]2
  • Analysis of 2,3-dihydro-2,2-dimethyl-7-benzofuranol in carbofuran by variable-temperature control system using liquid chromatography has been reported.2,3-dihydro-2,2-dimethyl-7-benzofuranol was used as starting reagent in the preparation of aminoalkanol derivatives of 2,3-dihydro-2,2-dimethyl-7-benzofuranol

Specifications

CAS 57196-62-0
Molecular Formula C10H14ClNO
Linear Formula C10H14ClNO
MDL Number MFCD01717328
Quantity 100 mL
Molecular Weight (g/mol) 199.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.