Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene 98.0+%, TCI America™
SDP

Catalog No. M23455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23455G 5 g
M234525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23455G Supplier TCI America Supplier No. M23455G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63675-74-1
Molecular Formula C16H14O2S
Molecular Weight (g/mol) 270.35
MDL Number MFCD02083138
InChI Key HRWAGCVMOGWQJF-UHFFFAOYSA-N
PubChem CID 850941
IUPAC Name 6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene
SMILES COC1=CC=C(C=C1)C1=CC2=CC=C(OC)C=C2S1

Specifications

Melting Point 198°C
Quantity 5 g
Formula Weight 270.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.