Learn More
Medchemexpress LLC 6-methoxy-2-(5-methyl-2-furanyl)-N-(1-methyl-4-piperidinyl)-7-[3-(1-pyrrolidinyl)propoxy]-4-quinolin | 1846570-31-7 | 98.7% | 478.63 g/mol | C28H38N4O3 | 50 MG
SDP
Catalog No. 5000248906
Encompass_Preferred
CM-272 is a reversible small-molecule inhibitor that selectively targets the histone methyltransferase G9a and DNA methyltransferases (DNMTs), showing antiproliferative and pro-apoptotic activity in preclinical cancer models and inducing immunogenic responses.
  • Dual inhibition of G9a and DNA methyltransferases enables epigenetic modulation.
  • Low-nanomolar potency against primary target with measurable activity on DNMT isoforms.
  • Demonstrated antiproliferative and pro-apoptotic effects in cellular and animal studies.
  • High chemical purity suitable for research applications.
  • Available in small milligram quantities for in vitro and in vivo experiments.
Catalog No. 50-002-48906 Supplier Medchemexpress LLC Supplier No. HY10192550MG
CM-272 is a reversible small-molecule inhibitor that selectively targets the histone methyltransferase G9a and DNA methyltransferases (DNMTs), showing antiproliferative and pro-apoptotic activity in preclinical cancer models and inducing immunogenic responses.
  • Dual inhibition of G9a and DNA methyltransferases enables epigenetic modulation.
  • Low-nanomolar potency against primary target with measurable activity on DNMT isoforms.
  • Demonstrated antiproliferative and pro-apoptotic effects in cellular and animal studies.
  • High chemical purity suitable for research applications.
  • Available in small milligram quantities for in vitro and in vivo experiments.