Learn More
Cayman Chemical 6-methoxy-2-Naphthoic Acid, Purity: ≥98%, CAS: 2471-70-7, 10 g
SDP
Catalog No. 502913368
Encompass_Preferred
An active metabolite of nabumetone; inhibits COX-1 and LPS-induced COX-2 activities in isolated human whole blood (IC50s = 31.01 and 19.84 µM, respectively); a potential impurity in commercial preparations of naproxen
Catalog No. 50-291-3368 Supplier Cayman Chemical Supplier No. 3537410 G
An active metabolite of nabumetone; inhibits COX-1 and LPS-induced COX-2 activities in isolated human whole blood (IC50s = 31.01 and 19.84 µM, respectively); a potential impurity in commercial preparations of naproxen