Learn More
Medchemexpress LLC 6-Methoxy-2-naphthoic acid | 2471-70-7 | 99.7% | 1 ML
SDP
Catalog No. 5000410759
Encompass_Preferred
6-Methoxy-2-naphthoic acid (6-MNA) is the active metabolite of Nabumetone. It functions as an inhibitor of COX-1 and COX-2 enzymes and inhibits the synthesis of gastric mucosal prostaglandin E2. This compound is valuable in research related to inflammation and pain-related diseases, and it also serves as an impurity in Naproxen and a building block for synthesizing other active compounds.
  • Active metabolite of Nabumetone
  • Inhibitor of COX-1 and COX-2
  • Inhibits gastric mucosal prostaglandin E2 synthesis
  • Used in inflammation and pain research
  • Impurity in Naproxen
  • Precursor for active compound synthesis
Catalog No. 50-004-10759 Supplier Medchemexpress LLC Supplier No. HYB212110MM/1ML
6-Methoxy-2-naphthoic acid (6-MNA) is the active metabolite of Nabumetone. It functions as an inhibitor of COX-1 and COX-2 enzymes and inhibits the synthesis of gastric mucosal prostaglandin E2. This compound is valuable in research related to inflammation and pain-related diseases, and it also serves as an impurity in Naproxen and a building block for synthesizing other active compounds.
  • Active metabolite of Nabumetone
  • Inhibitor of COX-1 and COX-2
  • Inhibits gastric mucosal prostaglandin E2 synthesis
  • Used in inflammation and pain research
  • Impurity in Naproxen
  • Precursor for active compound synthesis