Learn More
Medchemexpress LLC 6-Methoxy-2-naphthoic acid | 2471-70-7 | 99.7% | 5 G
SDP
Catalog No. 5000411024
Encompass_Preferred
6-Methoxy-2-naphthoic acid (6-MNA) is the active metabolite of Nabumetone and an inhibitor of COX-1 and COX-2. It inhibits the synthesis of gastric mucosal prostaglandin E2. This compound is used in research related to inflammation and pain-related diseases, and it is also an impurity found in Naproxen. It can be used in the synthesis of other active compounds.
Catalog No. 50-004-11024 Supplier Medchemexpress LLC Supplier No. HYB21215G
Add to Cart
Add to Cart
6-Methoxy-2-naphthoic acid (6-MNA) is the active metabolite of Nabumetone and an inhibitor of COX-1 and COX-2. It inhibits the synthesis of gastric mucosal prostaglandin E2. This compound is used in research related to inflammation and pain-related diseases, and it is also an impurity found in Naproxen. It can be used in the synthesis of other active compounds.