Learn More
Medchemexpress LLC 6-Methoxy-7-(3-(pyrrolidin-1-yl)propoxy)-4-(tetrahydro-2H-pyran-4-yl)quinolin-2-amine | 99.4% | 385.50 | 25 MG
SDP
Catalog No. 5000394880
Encompass_Preferred
MS152 is an orally bioactive inhibitor of EHMT2/G9a. It reactivates maternally silenced Prader-Willi syndrome (PWS) genes in brain and liver tissues of PWS mouse models. This product is for research use only.
  • Oral bioactive inhibitor of EHMT2/G9a.
  • Reactivates maternally silenced Prader-Willi syndrome (PWS) genes.
  • Suitable for research applications.
Catalog No. 50-003-94880 Supplier Medchemexpress LLC Supplier No. 25MGHY161759
MS152 is an orally bioactive inhibitor of EHMT2/G9a. It reactivates maternally silenced Prader-Willi syndrome (PWS) genes in brain and liver tissues of PWS mouse models. This product is for research use only.
  • Oral bioactive inhibitor of EHMT2/G9a.
  • Reactivates maternally silenced Prader-Willi syndrome (PWS) genes.
  • Suitable for research applications.