Learn More
Cayman Chemical 6-methoxy DMT, Purity: ≥98%, CAS: 2426-88-2, 5 mg
SDP
Catalog No. 502950186
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; does not induce generalized responses in rats trained to discriminate DOM from saline; intended for research and forensic applications
Catalog No. 50-295-0186 Supplier Cayman Chemical Supplier No. 336265 MG
An analytical reference standard categorized as a tryptamine; does not induce generalized responses in rats trained to discriminate DOM from saline; intended for research and forensic applications