Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Methoxyquinoline | 5263-87-6 | MFCD00006800 | 100.0% | 159.19 g/mol | C10H9NO | 5 G
SDP

Catalog No. 5000301862
Encompass_Preferred

6-Methoxyquinoline is an aromatic quinoline derivative used as a biochemical assay reagent and a synthetic intermediate. It is supplied with solubility and storage guidance for laboratory handling.

  • Aromatic quinoline derivative suitable for biochemical assays and organic synthesis.
  • Molecular formula C10H9NO; molecular weight 159.19 g/mol.
  • Reported purity 99.99% for research applications.
  • Soluble in DMSO at 200 mg/mL; sonication may be required.
  • Storage stability: 3 years at room temperature; in solvent, 2 years at -80°C and 1 year at -20°C.
  • CAS number 5263-87-6 for unambiguous identification.

Catalog No. 50-003-01862 Supplier Medchemexpress LLC Supplier No. HYW0172325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Methoxyquinoline is an aromatic quinoline derivative used as a biochemical assay reagent and a synthetic intermediate. It is supplied with solubility and storage guidance for laboratory handling.

  • Aromatic quinoline derivative suitable for biochemical assays and organic synthesis.
  • Molecular formula C10H9NO; molecular weight 159.19 g/mol.
  • Reported purity 99.99% for research applications.
  • Soluble in DMSO at 200 mg/mL; sonication may be required.
  • Storage stability: 3 years at room temperature; in solvent, 2 years at -80°C and 1 year at -20°C.
  • CAS number 5263-87-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.