Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-methoxyquinoline (p-quinanisole) | 5263-87-6 | MFCD00006800 | 99.7% | 159.19 | C10H9NO | 25 G
SDP

Catalog No. 5000304208
Encompass_Preferred

6-Methoxyquinoline (p-quinanisole) is a quinoline-derived laboratory reagent used as a building block in organic synthesis and as a reagent in biochemical assays. It is supplied with documentation for research use and is characterized for purity and storage stability.

  • High purity (≈99.7%) suitable for research applications.
  • Molecular formula C10H9NO; molecular weight 159.19.
  • Available in multiple pack sizes, including 25 g.
  • Appearance: off-white to yellow; solid below ~18°C, liquid above ~20°C.
  • Supplied with datasheet, COA, SDS, and handling instructions.
  • Storage: product stable at room temperature; stock solutions stable at -20°C to -80°C.

Catalog No. 50-003-04208 Supplier Medchemexpress LLC Supplier No. 25GHYW017232
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Methoxyquinoline (p-quinanisole) is a quinoline-derived laboratory reagent used as a building block in organic synthesis and as a reagent in biochemical assays. It is supplied with documentation for research use and is characterized for purity and storage stability.

  • High purity (≈99.7%) suitable for research applications.
  • Molecular formula C10H9NO; molecular weight 159.19.
  • Available in multiple pack sizes, including 25 g.
  • Appearance: off-white to yellow; solid below ~18°C, liquid above ~20°C.
  • Supplied with datasheet, COA, SDS, and handling instructions.
  • Storage: product stable at room temperature; stock solutions stable at -20°C to -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.