Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-methyl-1,4-naphthoquinone | 605-93-6 | MFCD04039611 | 98.5% | 172.18 g·mol⁻1 | C11H8O2 | 25 MG
SDP

Catalog No. 5000284228
Encompass_Preferred

6-Methyl-1,4-naphthoquinone is a methyl-substituted naphthoquinone research reagent used in biochemical assays and small-molecule synthesis. It is supplied as a laboratory-scale solid and intended for research use only; consult the product data sheet and COA for handling, storage, and batch-specific information.

  • High purity (98.49%) for reproducible experimental results.
  • Molecular weight 172.18 g·mol⁻1 and formula C11H8O2.
  • Available in multiple laboratory pack sizes, for example 25 mg, 50 mg, 100 mg.
  • Solid, yellow to brown appearance for easy visual identification.
  • CAS number 605-93-6 for unambiguous substance identification.
  • Data sheet and certificate of analysis available for batch-specific details.

Catalog No. 50-002-84228 Supplier Medchemexpress LLC Supplier No. HYW58594325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Methyl-1,4-naphthoquinone is a methyl-substituted naphthoquinone research reagent used in biochemical assays and small-molecule synthesis. It is supplied as a laboratory-scale solid and intended for research use only; consult the product data sheet and COA for handling, storage, and batch-specific information.

  • High purity (98.49%) for reproducible experimental results.
  • Molecular weight 172.18 g·mol⁻1 and formula C11H8O2.
  • Available in multiple laboratory pack sizes, for example 25 mg, 50 mg, 100 mg.
  • Solid, yellow to brown appearance for easy visual identification.
  • CAS number 605-93-6 for unambiguous substance identification.
  • Data sheet and certificate of analysis available for batch-specific details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.