Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-methyl-1,4-naphthoquinone | 605-93-6 | MFCD04039611 | 98.5% | 172.18 g/mol | C11H8O2 | 100 MG
SDP

Catalog No. 5000284638
Encompass_Preferred

6-Methyl-1,4-naphthoquinone is a redox-active small-molecule reagent supplied as a purified solid for biochemical research and assay development. It is used in studies of naphthoquinone chemistry, redox biology, and small-molecule screening applications.

  • High purity (98.49%).
  • Chemical formula C11H8O2 and molecular weight 172.18 g/mol.
  • Available in multiple package sizes (25 mg, 50 mg, 100 mg, 500 mg; larger sizes on request).
  • Storage: powder at -20°C; in solvent store at -80°C for long-term, -20°C for short-term.
  • Includes supporting documents: data sheet, safety data sheet, and certificate of analysis.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-84638 Supplier Medchemexpress LLC Supplier No. HYW585943100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Methyl-1,4-naphthoquinone is a redox-active small-molecule reagent supplied as a purified solid for biochemical research and assay development. It is used in studies of naphthoquinone chemistry, redox biology, and small-molecule screening applications.

  • High purity (98.49%).
  • Chemical formula C11H8O2 and molecular weight 172.18 g/mol.
  • Available in multiple package sizes (25 mg, 50 mg, 100 mg, 500 mg; larger sizes on request).
  • Storage: powder at -20°C; in solvent store at -80°C for long-term, -20°C for short-term.
  • Includes supporting documents: data sheet, safety data sheet, and certificate of analysis.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.