Learn More
Medchemexpress LLC 6-Methyl-1H-indole | 3420-02-8 | 131.18 | 10 G
SDP
Catalog No. 5000460401
Encompass_Preferred
6-Methyl-1H-indole is a biochemical reagent used as a biological material or organic compound for life science research. It serves as a reactant in the preparation of various compounds for medicinal and biological applications.
  • Reactant for tryptophan dioxygenase inhibitors, which are potential anticancer immunomodulators
  • Reactant for indole-linked triazole derivatives as antifungal agents
  • Reactant for N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for managing hyperglycemia in diabetes
  • Reactant for aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents
  • Reactant for indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors
  • Reactant for arylsulfonylacetylindoles as anti-HIV-1 agents
  • Used in the synthesis of benz[c,d]indol-3(1H)-one derivatives
Catalog No. 50-004-60401 Supplier Medchemexpress LLC Supplier No. HYW00734910G
6-Methyl-1H-indole is a biochemical reagent used as a biological material or organic compound for life science research. It serves as a reactant in the preparation of various compounds for medicinal and biological applications.
  • Reactant for tryptophan dioxygenase inhibitors, which are potential anticancer immunomodulators
  • Reactant for indole-linked triazole derivatives as antifungal agents
  • Reactant for N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for managing hyperglycemia in diabetes
  • Reactant for aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents
  • Reactant for indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors
  • Reactant for arylsulfonylacetylindoles as anti-HIV-1 agents
  • Used in the synthesis of benz[c,d]indol-3(1H)-one derivatives