Learn More
Medchemexpress LLC 6-Methyl-1H-indole | 3420-02-8 | 131.18 | 25 G
SDP
Catalog No. 5000449326
Encompass_Preferred
6-Methyl-1H-indole is a biochemical reagent used as a biological material or organic compound for life science related research. It serves as a reactant for the preparation of various compounds with diverse applications.
  • Reactant for preparation of tryptophan dioxygenase inhibitors
  • Reactant for preparation of indole linked triazole derivatives
  • Reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors
  • Reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles
  • Reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors
  • Reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents
  • Used in the synthesis of benz[c,d]indol-3(1H)-one derivatives
Catalog No. 50-004-49326 Supplier Medchemexpress LLC Supplier No. 25GHYW007349
6-Methyl-1H-indole is a biochemical reagent used as a biological material or organic compound for life science related research. It serves as a reactant for the preparation of various compounds with diverse applications.
  • Reactant for preparation of tryptophan dioxygenase inhibitors
  • Reactant for preparation of indole linked triazole derivatives
  • Reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors
  • Reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles
  • Reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors
  • Reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents
  • Used in the synthesis of benz[c,d]indol-3(1H)-one derivatives