Learn More
Medchemexpress LLC 6-methyl-2-(((3-phenoxypropyl)amino)methyl)quinolin-4-ol | 1434515-70-4 | 99.5% | 322.40 g/mol | C20H22N2O2 | 10MG
SDP
Catalog No. 5000236378
Encompass_Preferred
UCSF924 is a selective dopamine D4 receptor (DRD4) partial agonist chemical probe with nanomolar potency (EC50 = 4.2 nM) and exceptional selectivity against other GPCRs. It is supplied as a white to off-white solid with high purity for use in receptor pharmacology and neuropsychiatric research.
  • Selective DRD4 partial agonist with EC50 = 4.2 nM.
  • High selectivity against more than 320 non-olfactory GPCRs.
  • High purity: 99.5%.
  • White to off-white solid suitable for laboratory use.
  • Appropriate for cell-based assays and receptor pharmacology studies.
Catalog No. 50-002-36378 Supplier Medchemexpress LLC Supplier No. HY12575110MG
UCSF924 is a selective dopamine D4 receptor (DRD4) partial agonist chemical probe with nanomolar potency (EC50 = 4.2 nM) and exceptional selectivity against other GPCRs. It is supplied as a white to off-white solid with high purity for use in receptor pharmacology and neuropsychiatric research.
  • Selective DRD4 partial agonist with EC50 = 4.2 nM.
  • High selectivity against more than 320 non-olfactory GPCRs.
  • High purity: 99.5%.
  • White to off-white solid suitable for laboratory use.
  • Appropriate for cell-based assays and receptor pharmacology studies.