Learn More
Medchemexpress LLC 6-methyl-2-[(3-phenoxypropyl)aminomethyl]quinolin-4(1H)-one | 1434515-70-4 | 99.5% | 322.40 g·mol-1 | C20H22N2O2 | 100MG
SDP
Catalog No. 5000239167
Encompass_Preferred
UCSF924 is a potent and selective dopamine D4 receptor (DRD4) partial agonist used as a chemical probe in neuropsychiatric and pharmacology research. It exhibits an EC50 of 4.2 nM and a Ki of approximately 3 nM for DRD4, with reported lack of off-target activity against more than 320 non-olfactory GPCRs. Supplied as a high-purity research compound for biochemical and cellular studies. Not for human or veterinary use.
  • Potent DRD4 partial agonist (EC50 4.2 nM).
  • High affinity for DRD4 (Ki ≈ 3 nM).
  • Demonstrated selectivity with minimal off-target GPCR activity.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple packaging formats and DMSO solutions for assay use.
Catalog No. 50-002-39167 Supplier Medchemexpress LLC Supplier No. HY125751100MG
UCSF924 is a potent and selective dopamine D4 receptor (DRD4) partial agonist used as a chemical probe in neuropsychiatric and pharmacology research. It exhibits an EC50 of 4.2 nM and a Ki of approximately 3 nM for DRD4, with reported lack of off-target activity against more than 320 non-olfactory GPCRs. Supplied as a high-purity research compound for biochemical and cellular studies. Not for human or veterinary use.
  • Potent DRD4 partial agonist (EC50 4.2 nM).
  • High affinity for DRD4 (Ki ≈ 3 nM).
  • Demonstrated selectivity with minimal off-target GPCR activity.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple packaging formats and DMSO solutions for assay use.