Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-methyl-2-((3-phenoxypropylamino)methyl)quinolin-4(1H)-one | 1434515-70-4 | 99.5% | 322.40 | C20H22N2O2 | 50MG
SDP

Catalog No. 5000237846
Encompass_Preferred

UCSF924 is a potent and selective dopamine D4 receptor (DRD4) partial agonist used as a chemical probe in receptor pharmacology and neuropsychiatric research. It exhibits low-nanomolar activity (EC50 4.2 nM) and is supplied as a high-purity solid with recommended low-temperature storage for long-term stability.

  • Potent DRD4 partial agonist (EC50 4.2 nM).
  • High selectivity against other G protein-coupled receptors.
  • Reported purity 99.53%.
  • Molecular formula C20H22N2O2; molecular weight 322.40.
  • Available in multiple pack sizes including 50 mg.
  • Appearance: white to off-white solid; powder storage recommended at -20°C for long-term stability.

Catalog No. 50-002-37846 Supplier Medchemexpress LLC Supplier No. HY12575150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UCSF924 is a potent and selective dopamine D4 receptor (DRD4) partial agonist used as a chemical probe in receptor pharmacology and neuropsychiatric research. It exhibits low-nanomolar activity (EC50 4.2 nM) and is supplied as a high-purity solid with recommended low-temperature storage for long-term stability.

  • Potent DRD4 partial agonist (EC50 4.2 nM).
  • High selectivity against other G protein-coupled receptors.
  • Reported purity 99.53%.
  • Molecular formula C20H22N2O2; molecular weight 322.40.
  • Available in multiple pack sizes including 50 mg.
  • Appearance: white to off-white solid; powder storage recommended at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.