Learn More
Medchemexpress LLC 6-methyl-2-((3-phenoxypropylamino)methyl)quinolin-4(1H)-one | 1434515-70-4 | 99.5% | 322.40 g/mol | C20H22N2O2 | 25MG
SDP
Catalog No. 5000235638
Encompass_Preferred
UCSF924 is a potent, selective dopamine D4 receptor (DRD4) partial agonist research probe for neuropsychiatric and GPCR pharmacology studies. It exhibits an EC50 of 4.2 nM at DRD4 and shows no significant off-target activity across more than 320 non-olfactory GPCRs. The compound is provided as a solid with high purity suitable for in vitro assays.
  • High potency at DRD4 (EC50 4.2 nM).
  • Exceptional selectivity versus non-olfactory GPCRs (>320 tested).
  • High purity (99.5%).
  • Molecular weight 322.40 g/mol and formula C20H22N2O2.
  • Available as a solid and as a 10 mM solution in DMSO for convenient use.
  • Suitable for cell-based pharmacology and selectivity studies.
Catalog No. 50-002-35638 Supplier Medchemexpress LLC Supplier No. HY12575125MG
UCSF924 is a potent, selective dopamine D4 receptor (DRD4) partial agonist research probe for neuropsychiatric and GPCR pharmacology studies. It exhibits an EC50 of 4.2 nM at DRD4 and shows no significant off-target activity across more than 320 non-olfactory GPCRs. The compound is provided as a solid with high purity suitable for in vitro assays.
  • High potency at DRD4 (EC50 4.2 nM).
  • Exceptional selectivity versus non-olfactory GPCRs (>320 tested).
  • High purity (99.5%).
  • Molecular weight 322.40 g/mol and formula C20H22N2O2.
  • Available as a solid and as a 10 mM solution in DMSO for convenient use.
  • Suitable for cell-based pharmacology and selectivity studies.