Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI 6-Methyl-2-thiouracil, >98.0% Purity, Powder, 25 g

Catalog No. 502780952
Encompass_Preferred

6-Methyl-2-thiouracil, also known as Methiocil, is a chemical substance used for laboratory research purposes. This compound is provided as a white to almost white powder or crystal.

  • Molecular formula: C5H6N2OS
  • Purity: ≥98.0% (T)
  • Molecular weight: 142.18
  • Physical state: Solid at 20°C
  • Storage: Room temperature (recommended in a cool and dark place, <15°C)

Catalog No. 50-278-0952 Supplier Grainger Supplier No. 19KY23
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Methyl-2-thiouracil, also known as Methiocil, is a chemical substance used for laboratory research purposes. This compound is provided as a white to almost white powder or crystal.

  • Molecular formula: C5H6N2OS
  • Purity: ≥98.0% (T)
  • Molecular weight: 142.18
  • Physical state: Solid at 20°C
  • Storage: Room temperature (recommended in a cool and dark place, <15°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.