Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-3,5-heptadiene-2-one, 96%, Thermo Scientific™

Catalog No. AC453211000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453211000 100 g
AC453210050 5 g
AC453210250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC453211000 Supplier Thermo Scientific Chemicals Supplier No. 453211000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1604-28-0
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
InChI Key KSKXSFZGARKWOW-GQCTYLIASA-N
Synonym 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
PubChem CID 5370101
IUPAC Name (3E)-6-methylhepta-3,5-dien-2-one
SMILES CC(=CC=CC(=O)C)C

Specifications

Density 0.8930g/mL
Boiling Point 168.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Linear Formula (CH3)2C=CHCH=CHCOCH3
Quantity 100 g
Specific Gravity 0.893
Formula Weight 124.18
Percent Purity 96%
Chemical Name or Material 6-Methyl-3,5-heptadiene-2-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 216-507-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.