Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine

Catalog No. SYX002601G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002601G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002601G Supplier Sigma Aldrich Supplier No. SYX002601G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Crotyl chloride reacts with trichlorosilane in the presence of tertiary amine and copper salts to give the corresponding allyltrichlorosilanes.crotyl chloride has been used:
  • To investihgate the kinetics of the reactions of atomic chlorine with series of unsaturated aldehydes and ketones at 298k, to probe structure-reactivity relationships
  • In the preparation of (e)-crotyltrichlorosilane

Specifications

CAS 7496-50-6
Molecular Formula C8H12N2S
Linear Formula C8H12N2S
MDL Number MFCD00805140
Quantity 100 mL
Molecular Weight (g/mol) 168.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.