Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-5-nitroquinoline, 97%, Thermo Scientific™

Catalog No. AAA1139314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1139314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1139314 Supplier Thermo Scientific Chemicals Supplier No. A1139314

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23141-61-9
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00024021
InChI Key CENBTULRDDPOGR-UHFFFAOYSA-N
Synonym 5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
PubChem CID 90009
IUPAC Name 6-methyl-5-nitroquinoline
SMILES CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]

Specifications

Melting Point 115°C to 118°C
Quantity 25 g
Beilstein 154688
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 6-Methyl-5-nitroquinoline
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.