Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylbenzimidazole-5-carboxylic acid hydrochloride, 97%

Catalog No. AAH2592603
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2592603 1 g
AAH2592609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2592603 Supplier Thermo Scientific Chemicals Supplier No. H2592603
Add to Cart

CAS: 10351-76-5 | C9H9ClN2O2 | 212.633 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10351-76-5
Molecular Formula C9H9ClN2O2
Molecular Weight (g/mol) 212.633
MDL Number MFCD09909565
InChI Key QCJPDBNTTAJIRH-UHFFFAOYSA-N
Synonym 6-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 6-methyl-5-benzimidazole carboxylic acid, 1h-benzimidazole-5-carboxylic acid,6-methyl, 1h-benzimidazole-5-carboxylicacid, 6-methyl, 1h-benzimidazole-6-carboxylicacid, 5-methyl, 5-methyl-3h-benzoimidazole-6-carboxylic acid, 5-methyl-1h-benzo d imidazole-6-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 6-methyl-9ci
PubChem CID 45925698
IUPAC Name 6-methyl-1H-benzimidazole-5-carboxylic acid;hydrochloride
SMILES CC1=CC2=C(C=C1C(=O)O)N=CN2.Cl

Specifications

Melting Point 315°C (decomposition)
Quantity 1 g
Formula Weight 212.64
Percent Purity 97%
Chemical Name or Material 6-Methylbenzimidazole-5-carboxylic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.