Learn More
Medchemexpress LLC HY-N6630 100mg Medchemexpress, 6-Methylflavone CAS:29976-75-8 Purity:>98%
SDP
Catalog No. 501933964
Encompass_Preferred
Medchemexpress, HY-N6630 100mg 6-Methylflavone CAS:29976-75-8 6-Methylflavone is an activator of α 1 β 2 γ 2L and α 1 β 2 GABA A receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3964 Supplier Medchemexpress LLC Supplier No. HYN6630100MG
Medchemexpress, HY-N6630 100mg 6-Methylflavone CAS:29976-75-8 6-Methylflavone is an activator of α 1 β 2 γ 2L and α 1 β 2 GABA A receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.