Learn More
Apexbio Technology LLC 6-METHYLFLAVONE-200MG
SDP
Catalog No. 5000568573
Encompass_Preferred
6-Methylflavone (CAS No 29976-75-8) is a synthetic flavonoid with potential therapeutic applications in biomedical research It is primarily studied for its interaction with cellular pathways including modulation of ion channels and neurotransmitter receptors Research indicates that it exhibits biological activity in the nanomolar to low micromolar range showcasing its efficacy in modulating physiological responses Common applications include its use in in vitro cell models and in vivo animal studies to investigate its potential in neuroprotection and anti-inflammatory effects 6-Methylflavone is also utilized in drug screening programs aiming to develop novel therapeutic agents Experimental concentrations typically depend on the specific research design allowing flexibility in various biological and pharmacological assays to elucidate its mechanism of action and therapeutic viability
Catalog No. 50-005-68573 Supplier Apexbio Technology LLC Supplier No. C6622200
6-Methylflavone (CAS No 29976-75-8) is a synthetic flavonoid with potential therapeutic applications in biomedical research It is primarily studied for its interaction with cellular pathways including modulation of ion channels and neurotransmitter receptors Research indicates that it exhibits biological activity in the nanomolar to low micromolar range showcasing its efficacy in modulating physiological responses Common applications include its use in in vitro cell models and in vivo animal studies to investigate its potential in neuroprotection and anti-inflammatory effects 6-Methylflavone is also utilized in drug screening programs aiming to develop novel therapeutic agents Experimental concentrations typically depend on the specific research design allowing flexibility in various biological and pharmacological assays to elucidate its mechanism of action and therapeutic viability