Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. AAL1750803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1750803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1750803 Supplier Thermo Scientific Chemicals Supplier No. L1750803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4771-49-7 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4771-49-7
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00049347
InChI Key LZERQSJGPXFAKB-UHFFFAOYSA-N
Synonym 6-methylindole-3-carboxaldehyde, 6-methylindole-3-carboxyaldehyde, 1h-indole-3-carboxaldehyde, 6-methyl, 6-methylindole-3-carbaldehyde, 6-methyl-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde,6-methyl, pubchem7241, acmc-209ka7, ksc497o2l, 6-methylindole-3-caboxaldehyde
PubChem CID 4777902
IUPAC Name 6-methyl-1H-indole-3-carbaldehyde
SMILES CC1=CC=C2C(NC=C2C=O)=C1

Specifications

Melting Point 185°C to 189°C
Quantity 1 g
Beilstein 386914
Sensitivity Air Sensitive
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 6-Methylindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.