Learn More
Cayman Chemical 6-Methylmercaptopurine, Purity: ≥95%, CAS: 50-66-8, 50 mg
SDP
Catalog No. 502950787
Encompass_Preferred
A metabolite of 6-MP; formed from 6-MP via methylation by thiopurine S-methyltransferase; levels are increased in patients with altered thiopurine metabolism and associated with therapeutic resistance to 6-MP
Catalog No. 50-295-0787 Supplier Cayman Chemical Supplier No. 3435650 MG
A metabolite of 6-MP; formed from 6-MP via methylation by thiopurine S-methyltransferase; levels are increased in patients with altered thiopurine metabolism and associated with therapeutic resistance to 6-MP